siRNA Details
| Gene Symbol: KDM6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080424.1 | NP_001073893.1 | 11 | 1715 | ||
| XM_005256549.3 | XP_005256606.1 | 12 | 1867 | |||
| XM_005256550.4 | XP_005256607.1 | 11 | 1709 | |||
| XM_005256551.3 | XP_005256608.1 | 11 | 1748 | |||
| XM_005256552.4 | XP_005256609.1 | 11 | 2721 | |||
| XM_006721483.3 | XP_006721546.1 | 12 | 1812 | |||
| XM_011523750.2 | XP_011522052.1 | 12 | 3072 | |||
| XM_011523752.2 | XP_011522054.1 | 11 | 1712 | |||
| XM_017024381.1 | XP_016879870.1 | 11 | 3158 | |||
| GenBank mRNA | AB002344.2 | - | 11 | 1720 | ||

KDM6B
Genomic Map