siRNA Details
| Gene Symbol: PLEKHM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015164.2 | NP_055979.2 | 9 | 1653 | ||
| XM_005245790.3 | XP_005245847.1 | 8 | 1788 | |||
| XM_005245791.4 | XP_005245848.1 | 4 | 921 | |||
| XM_017000757.1 | XP_016856246.1 | 9 | 1560 | |||
| XM_017000758.1 | XP_016856247.1 | 8 | 1500 | |||
| GenBank mRNA | AB020649.1 | - | 9 | 1429 | ||
| BC016488.1 | - | 1 | 335 | |||
| BC040441.1 | - | 9 | 1656 | |||

PLEKHM2
Genomic Map