siRNA Details
| Gene Symbol: PLEKHM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015164.2 | NP_055979.2 | 9 | 1703 | ||
| XM_005245790.3 | XP_005245847.1 | 8 | 1838 | |||
| XM_005245791.4 | XP_005245848.1 | 4 | 971 | |||
| XM_017000757.1 | XP_016856246.1 | 9 | 1610 | |||
| XM_017000758.1 | XP_016856247.1 | 8 | 1550 | |||
| GenBank mRNA | AB020649.1 | - | 9 | 1479 | ||
| BC016488.1 | - | 1 | 385 | |||
| BC040441.1 | - | 9 | 1706 | |||
| BC042103.1 | - | 1 | 33 | |||
| BC068599.1 | - | 1 | 58 | |||

PLEKHM2
Genomic Map