siRNA Details
| Gene Symbol: NBEAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015175.2 | NP_055990.1 | 27 | 4271 | ||
| XM_006713072.3 | XP_006713135.1 | 26 | 4187 | |||
| XM_011533533.2 | XP_011531835.1 | 27 | 4268 | |||
| XM_017006010.1 | XP_016861499.1 | 27 | 4268 | |||
| XM_017006011.1 | XP_016861500.1 | 27 | 4124 | |||
| XM_017006012.1 | XP_016861501.1 | 26 | 4187 | |||
| XM_017006013.1 | XP_016861502.1 | 27 | 4268 | |||
| XM_017006014.1 | XP_016861503.1 | 26 | 4043 | |||
| XM_017006015.1 | XP_016861504.1 | 25 | 3899 | |||
| XM_017006016.1 | XP_016861505.1 | 22 | 4845 | |||
| XM_017006017.1 | XP_016861506.1 | 4 | 687 | |||
| GenBank mRNA | AK131104.1 | - | 27 | 4111 | ||

NBEAL2
Genomic Map