siRNA Details
| Gene Symbol: DTX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300727.1 | NP_001287656.1 | 8, 9 | 1466 | ||
| NM_015177.1 | NP_055992.1 | 8, 9 | 1875 | |||
| XM_006718483.3 | XP_006718546.1 | 8, 9 | 1919 | |||
| GenBank mRNA | AB023154.1 | - | 8, 9 | 1722 | ||
| BC119011.1 | - | 8, 9 | 1466 | 40085251 | ||
| BC122861.1 | - | 8, 9 | 1466 | 40085252 | ||

DTX4
Genomic Map