siRNA Details
| Gene Symbol: ELAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165962.1 | NP_001159434.1 | 23 | 2747 | ||
| NM_018127.6 | NP_060597.4 | 24 | 2867 | |||
| NM_173717.1 | NP_776065.1 | 24 | 2864 | |||
| GenBank mRNA | AF304370.1 | - | 24 | 2750 | ||
| AK001392.1 | - | 24 | 2820 | |||
| AK074244.1 | - | 16 | 1989 | |||
| AK094012.1 | - | 9 | 2637 | |||
| AK094333.1 | - | 16 | 2578 | |||
| AK094687.1 | - | 24 | 2751 | |||
| AK124838.1 | - | 13 | 2296 | |||
| AK125030.1 | - | 22 | 3580 | |||
| AK300011.1 | - | 24 | 2742 | |||
| BC001939.1 | - | 24 | 2810 | 2820640 | ||
| BC004158.2 | - | 24 | 2810 | 2820640 | ||

ELAC2
Genomic Map