siRNA Details
| Gene Symbol: TBC1D12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015188.1 | NP_056003.1 | 13 | 4284 | ||
| XM_006717732.3 | XP_006717795.2 | 12 | 4203 | |||
| XM_006717733.2 | XP_006717796.1 | 14 | 3244 | |||
| XM_011539558.2 | XP_011537860.1 | 13 | 3295 | |||
| XM_017015996.1 | XP_016871485.1 | 11 | 4059 | |||
| XM_017015998.1 | XP_016871487.1 | 12 | 3071 | |||
| XM_017015999.1 | XP_016871488.1 | 12 | 3287 | |||
| GenBank mRNA | AJ404330.1 | - | 1 | 836 | ||
| BC064323.1 | - | 4 | 2302 | |||

TBC1D12
Genomic Map