siRNA Details
| Gene Symbol: KIAA1107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000800.1 | XP_016856289.1 | 7 | 1730 | ||
| GenBank mRNA | AB029030.1 | - | 6 | 1501 | ||
| AK302496.1 | - | 6 | 1606 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA1107| Gene Symbol: KIAA1107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000800.1 | XP_016856289.1 | 7 | 1730 | ||
| GenBank mRNA | AB029030.1 | - | 6 | 1501 | ||
| AK302496.1 | - | 6 | 1606 | |||