siRNA Details
| Gene Symbol: KIAA1107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000800.1 | XP_016856289.1 | 2 | 124 | ||
| GenBank mRNA | AB029030.1 | - | 2 | 82 | ||
| AF017090.1 | - | 2 | 129 | |||
| AK302496.1 | - | 2 | 187 | |||
| DC331933.1 | - | 2 | 149 | |||

KIAA1107
Genomic Map