siRNA Details
| Gene Symbol: FKBP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015258.1 | NP_056073.1 | 28 | 4294 | ||
| XM_006717018.2 | XP_006717081.1 | 28 | 4181 | |||
| XM_006717019.1 | XP_006717082.1 | 27 | 4142 | |||
| XM_017014552.1 | XP_016870041.1 | 27 | 4112 | |||
| GenBank mRNA | AB014574.1 | - | 28 | 4221 | ||
| BC026048.1 | - | 3 | 1168 | |||
| BC077732.1 | - | 12 | 2504 | |||

FKBP15
Genomic Map