siRNA Details
| Gene Symbol: DPY19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015283.1 | NP_056098.1 | 22 | 4515 | ||
| XM_011515245.2 | XP_011513547.1 | 22 | 4664 | |||
| XM_011515246.2 | XP_011513548.1 | 21 | 4577 | |||
| GenBank mRNA | AB020684.1 | - | 19 | 4087 | ||
| BC050318.1 | - | 1 | 1856 | |||
| DQ287932.1 | - | 22 | 4515 | |||

DPY19L1
Genomic Map