siRNA Details
| Gene Symbol: U2SURP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080415.1 | NP_001073884.1 | 6, 7 | 667 | ||
| NM_001320219.1 | NP_001307148.1 | 6, 7 | 667 | |||
| NM_001320220.1 | NP_001307149.1 | 6, 7 | 667 | |||
| NM_001320222.1 | NP_001307151.1 | 6, 7 | 667 | |||
| XM_005247248.4 | XP_005247305.1 | 6, 7 | 673 | |||
| XM_017006038.1 | XP_016861527.1 | 6, 7 | 606 | |||
| GenBank mRNA | AB002330.1 | - | 6, 7 | 566 | ||
| AK296440.1 | - | 6, 7 | 587 | |||
| BC006474.1 | - | 6, 7 | 566 | |||
| BC111692.1 | - | 6, 7 | 1038 | 40028600 | ||

U2SURP
Genomic Map