siRNA Details
| Gene Symbol: ZNF629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080417.1 | NP_001073886.1 | 3 | 384 | ||
| XM_005255222.3 | XP_005255279.1 | 3 | 523 | |||
| GenBank mRNA | AB002324.1 | - | 3 | 350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF629| Gene Symbol: ZNF629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080417.1 | NP_001073886.1 | 3 | 384 | ||
| XM_005255222.3 | XP_005255279.1 | 3 | 523 | |||
| GenBank mRNA | AB002324.1 | - | 3 | 350 | ||