siRNA Details
| Gene Symbol: TMEM131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015348.1 | NP_056163.1 | 31 | 3902 | ||
| XM_005263909.1 | XP_005263966.1 | 32 | 3988 | |||
| XM_005263910.1 | XP_005263967.1 | 32 | 3988 | |||
| XM_005263911.1 | XP_005263968.1 | 31 | 3871 | |||
| XM_005263912.3 | XP_005263969.1 | 30 | 4076 | |||
| XM_011510869.1 | XP_011509171.1 | 31 | 3802 | |||
| GenBank mRNA | AU132630.1 | - | 2 | 169 | ||
| D87446.1 | - | 30 | 3440 | |||

TMEM131
Genomic Map