siRNA Details
| Gene Symbol: TMEM131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015348.1 | NP_056163.1 | 36 | 4994 | ||
| XM_005263909.1 | XP_005263966.1 | 37 | 5080 | |||
| XM_005263910.1 | XP_005263967.1 | 37 | 5080 | |||
| XM_005263911.1 | XP_005263968.1 | 36 | 4963 | |||
| XM_005263912.3 | XP_005263969.1 | 35 | 5168 | |||
| XM_011510869.1 | XP_011509171.1 | 36 | 4894 | |||
| GenBank mRNA | AK025852.1 | - | 6 | 642 | ||
| AK098792.1 | - | 5 | 817 | |||
| BX647728.1 | - | 1 | 3159 | |||
| D87446.1 | - | 35 | 4532 | |||

TMEM131
Genomic Map