siRNA Details
| Gene Symbol: EHBP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099409.1 | NP_001092879.1 | 9 | 1355 | ||
| XM_005273867.3 | XP_005273924.1 | 10 | 1394 | |||
| XM_005273868.3 | XP_005273925.1 | 10 | 1393 | |||
| XM_006718486.3 | XP_006718549.1 | 10 | 1392 | |||
| XM_006718487.3 | XP_006718550.1 | 10 | 1392 | |||
| XM_017017467.1 | XP_016872956.1 | 10 | 1142 | |||
| XM_017017468.1 | XP_016872957.1 | 10 | 1119 | |||
| GenBank mRNA | AK024451.1 | - | 5 | 767 | ||
| AL713799.1 | - | 1 | 239 | |||
| AL834433.1 | - | 9 | 1052 | |||

EHBP1L1
Genomic Map