siRNA Details
| Gene Symbol: TBC1D26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK292544.1 | - | 9 | 1067 | ||
| AK315847.1 | - | 9 | 1067 | |||
| BC028414.1 | - | 8 | 1637 | |||
| Gene Symbol: TBC1D28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039397.2 | NP_001034486.2 | 10 | 1234 | ||
| GenBank mRNA | AK097563.1 | - | 9 | 1522 | ||
| AL137747.1 | - | 1 | 61 | |||

TBC1D26
Genomic Map