siRNA Details
| Gene Symbol: TBC1D28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039397.2 | NP_001034486.2 | 10 | 1149 | ||
| GenBank mRNA | AK097563.1 | - | 9 | 1437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBC1D28| Gene Symbol: TBC1D28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039397.2 | NP_001034486.2 | 10 | 1149 | ||
| GenBank mRNA | AK097563.1 | - | 9 | 1437 | ||