siRNA Details
| Gene Symbol: C11orf86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136485.1 | NP_001129957.1 | 2 | 450 | ||
| GenBank mRNA | AK026328.1 | - | 2 | 456 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11orf86| Gene Symbol: C11orf86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136485.1 | NP_001129957.1 | 2 | 450 | ||
| GenBank mRNA | AK026328.1 | - | 2 | 456 | ||