siRNA Details
| Gene Symbol: LINC00879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015400.1 | 7, 8 | 932 | |||
| GenBank mRNA | BC038721.1 | - | 7, 8 | 935 | ||
| BC048119.1 | - | 7, 8 | 933 | |||
| BC104171.1 | - | 6, 7 | 767 | |||
| BC104172.1 | - | 4, 5 | 329 | |||
| BC115359.1 | - | 7, 8 | 851 | |||
| BC115360.1 | - | 7, 8 | 884 | |||

LINC00879
Genomic Map