siRNA Details
| Gene Symbol: LINC00879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015400.1 | 6 | 687 | |||
| GenBank mRNA | BC038721.1 | - | 6 | 690 | ||
| BC048119.1 | - | 6 | 688 | |||
| BC104171.1 | - | 5 | 522 | |||
| BC104172.1 | - | 3 | 160 | |||
| BC115359.1 | - | 6 | 606 | |||
| BC115360.1 | - | 6 | 639 | |||

LINC00879
Genomic Map