siRNA Details
| Gene Symbol: LINC01018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024424.2 | 1 | 270 | |||
| GenBank mRNA | BC106918.2 | - | 1 | 249 | ||
| BC106919.1 | - | 1 | 252 | |||
| BI602592.1 | - | 1 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01018| Gene Symbol: LINC01018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024424.2 | 1 | 270 | |||
| GenBank mRNA | BC106918.2 | - | 1 | 249 | ||
| BC106919.1 | - | 1 | 252 | |||
| BI602592.1 | - | 1 | 274 | |||