siRNA Details
| Gene Symbol: LOC255187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122044.1 | 1 | 334 | |||
| GenBank mRNA | AK056780.1 | - | 1 | 148 | ||
| BG178577.1 | - | 1 | 334 | |||
| BX106527.1 | - | 1 | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC255187| Gene Symbol: LOC255187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122044.1 | 1 | 334 | |||
| GenBank mRNA | AK056780.1 | - | 1 | 148 | ||
| BG178577.1 | - | 1 | 334 | |||
| BX106527.1 | - | 1 | 196 | |||