siRNA Details
| Gene Symbol: INTS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018142.2 | NP_060612.2 | 9 | 1262 | ||
| XM_005273556.3 | XP_005273613.1 | 9 | 1304 | |||
| XM_017013608.1 | XP_016869097.1 | 8 | 915 | |||
| XM_017013609.1 | XP_016869098.1 | 8 | 1239 | |||
| GenBank mRNA | AK001431.1 | - | 9 | 1182 | ||
| BC001915.1 | - | 7 | 899 | |||
| BC006209.2 | - | 7 | 899 | |||
| BC051282.1 | - | 7 | 913 | |||
| CR457262.1 | - | 6 | 792 | |||

INTS10
Genomic Map