siRNA Details
| Gene Symbol: LINC01619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046159.2 | 6 | 1875 | |||
| NR_046160.2 | 6 | 2037 | ||||
| NR_132339.1 | 5 | 2113 | ||||
| NR_132340.1 | 5 | 1962 | ||||
| NR_132341.1 | 5 | 1985 | ||||
| GenBank mRNA | AF086029.1 | - | 1 | 80 | ||
| AK055439.1 | - | 5 | 1953 | |||
| BC029844.1 | - | 6 | 1875 | |||

LINC01619
Genomic Map