siRNA Details
| Gene Symbol: SERPINA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080451.1 | NP_001073920.1 | 4 | 1066 | ||
| XM_006720105.3 | XP_006720168.1 | 4 | 621 | |||
| GenBank mRNA | BC146962.1 | - | 4 | 1030 | 9056890 | |
| BX248259.1 | - | 4 | 1062 | |||

SERPINA11
Genomic Map