siRNA Details
| Gene Symbol: CDCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005273733.3 | XP_005273790.1 | 8 | 1119 | ||
| XM_005273734.3 | XP_005273791.1 | 8 | 1119 | |||
| XM_011544743.2 | XP_011543045.1 | 8 | 1119 | |||
| XM_011544747.2 | XP_011543049.1 | 7 | 675 | |||
| XM_011544748.2 | XP_011543050.1 | 8 | 1144 | |||
| XM_017017158.1 | XP_016872647.1 | 8 | 1119 | |||
| Gene Symbol: LOC256676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB231703.1 | - | 1 | 55 | ||

CDCA5
Genomic Map