siRNA Details
| Gene Symbol: HSPA12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025015.2 | NP_079291.2 | 12 | 4528 | ||
| XM_005269672.3 | XP_005269729.1 | 13 | 4678 | |||
| XM_005269673.4 | XP_005269730.1 | 13 | 5622 | |||
| XM_011539579.2 | XP_011537881.1 | 14 | 4739 | |||
| XM_011539580.2 | XP_011537882.1 | 13 | 4684 | |||
| XM_017016032.1 | XP_016871521.1 | 10 | 4250 | |||
| GenBank mRNA | AB007877.1 | - | 12 | 4528 | ||

HSPA12A
Genomic Map