siRNA Details
| Gene Symbol: C20orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009984.2 | NP_001009984.1 | 17 | 1509 | ||
| XM_005260684.3 | XP_005260741.1 | 17 | 1503 | |||
| XM_017027783.1 | XP_016883272.1 | 14 | 1130 | |||
| GenBank mRNA | AK226100.1 | - | 12 | 933 | ||
| AK303375.1 | - | 8 | 878 | |||
| AL110249.1 | - | 8 | 474 | |||
| BC122529.1 | - | 8 | 727 | 40012183 | ||

C20orf194
Genomic Map