siRNA Details
| Gene Symbol: UBXN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015562.1 | NP_056377.1 | 11 | 2909 | ||
| XM_011512670.2 | XP_011510972.1 | 11 | 2857 | |||
| XM_011512671.2 | XP_011510973.1 | 10 | 2714 | |||
| GenBank mRNA | AB018337.1 | - | 11 | 2835 | ||
| AK129880.1 | - | 8 | 1505 | |||
| BC028986.1 | - | 10 | 2742 | |||

UBXN7
Genomic Map