siRNA Details
Gene Symbol: FAM198A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129908.2 | NP_001123380.2 | 4 | 1863 | ||
GenBank mRNA | AK091001.1 | - | 4 | 1793 | ||
AL117530.1 | - | 3 | 1106 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM198A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129908.2 | NP_001123380.2 | 4 | 1863 | ||
GenBank mRNA | AK091001.1 | - | 4 | 1793 | ||
AL117530.1 | - | 3 | 1106 |