siRNA Details
| Gene Symbol: INTS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080453.2 | NP_001073922.2 | 28 | 3879 | ||
| XM_011515260.1 | XP_011513562.1 | 28 | 3879 | |||
| XM_011515262.2 | XP_011513564.1 | 28 | 3920 | |||
| XM_017011959.1 | XP_016867448.1 | 28 | 3879 | |||
| XM_017011960.1 | XP_016867449.1 | 28 | 3879 | |||
| GenBank mRNA | AB037861.1 | - | 10 | 1339 | ||
| AY358482.1 | - | 3 | 340 | |||
| BC004286.1 | - | 9 | 1206 | |||

INTS1
Genomic Map