siRNA Details
| Gene Symbol: TMEM251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098621.1 | NP_001092091.1 | 1 | 282 | ||
| GenBank mRNA | CX785861.1 | - | 1 | 282 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM251| Gene Symbol: TMEM251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098621.1 | NP_001092091.1 | 1 | 282 | ||
| GenBank mRNA | CX785861.1 | - | 1 | 282 | ||