siRNA Details
| Gene Symbol: FAM184B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015688.1 | NP_056503.1 | 5 | 1502 | ||
| GenBank mRNA | AB033102.1 | - | 5 | 1315 | ||
| AK308144.1 | - | 5 | 1502 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM184B| Gene Symbol: FAM184B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015688.1 | NP_056503.1 | 5 | 1502 | ||
| GenBank mRNA | AB033102.1 | - | 5 | 1315 | ||
| AK308144.1 | - | 5 | 1502 | |||