siRNA Details
| Gene Symbol: PDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199898.1 | NP_001186827.1 | 5 | 604 | ||
| NM_001199899.1 | NP_001186828.1 | 4 | 403 | |||
| NM_001199900.1 | NP_001186829.1 | 4 | 566 | |||
| NM_002611.4 | NP_002602.2 | 4 | 566 | |||
| GenBank mRNA | AK055119.1 | - | 5 | 604 | ||
| AK290522.1 | - | 4 | 465 | |||
| AK293600.1 | - | 4 | 380 | |||
| BC005811.2 | - | 4 | 454 | 3831465 | ||
| BC040478.1 | - | 4 | 509 | 4690182 | ||
| BC063137.1 | - | 4 | 480 | 4272593 | ||
| BQ071206.1 | - | 465 | ||||
| DA199055.1 | - | 4 | 403 | |||

PDK2
Genomic Map