siRNA Details
| Gene Symbol: C12orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242696.1 | NP_001229625.1 | 4 | 888 | ||
| GenBank mRNA | BC038743.1 | - | 4 | 856 | ||
| BC117348.1 | - | 4 | 738 | |||
| BC126393.1 | - | 4 | 738 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12orf80| Gene Symbol: C12orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242696.1 | NP_001229625.1 | 4 | 888 | ||
| GenBank mRNA | BC038743.1 | - | 4 | 856 | ||
| BC117348.1 | - | 4 | 738 | |||
| BC126393.1 | - | 4 | 738 | |||