siRNA Details
| Gene Symbol: LOC283440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033958.1 | 5 | 542 | |||
| GenBank mRNA | BU657491.1 | - | 5 | 552 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 LOC283440
LOC283440| Gene Symbol: LOC283440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033958.1 | 5 | 542 | |||
| GenBank mRNA | BU657491.1 | - | 5 | 552 | ||
 Back To Top
 Back To Top