siRNA Details
| Gene Symbol: GXYLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099650.1 | NP_001093120.1 | 7 | 3461 | ||
| NM_173601.1 | NP_775872.1 | 8 | 3554 | |||
| XM_017019211.1 | XP_016874700.1 | 8 | 3293 | |||
| GenBank mRNA | AK021504.1 | - | 1 | 288 | ||
| AK021752.1 | - | 1 | 311 | |||
| AL832209.1 | - | 4 | 2691 | |||
| BC015597.2 | - | 4 | 2616 | |||

GXYLT1
Genomic Map