siRNA Details
| Gene Symbol: ASUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018164.2 | NP_060634.2 | 4 | 1017 | ||
| XM_005253422.3 | XP_005253479.1 | 4 | 976 | |||
| XM_011520750.2 | XP_011519052.1 | 4 | 550 | |||
| XM_011520751.2 | XP_011519053.1 | 4 | 976 | |||
| XM_011520752.2 | XP_011519054.1 | 4 | 976 | |||
| XM_017019632.1 | XP_016875121.1 | 4 | 550 | |||
| XM_017019633.1 | XP_016875122.1 | 3 | 942 | |||
| XM_017019634.1 | XP_016875123.1 | 3 | 942 | |||
| XM_017019635.1 | XP_016875124.1 | 4 | 550 | |||
| XM_017019636.1 | XP_016875125.1 | 4 | 976 | |||
| XM_017019637.1 | XP_016875126.1 | 1 | 109 | |||
| GenBank mRNA | AK001492.1 | - | 4 | 646 | ||
| AK001499.1 | - | 4 | 682 | |||
| AK225336.1 | - | 4 | 668 | |||
| AK297948.1 | - | 2 | 357 | |||
| BC003081.2 | - | 4 | 706 | 2989678 | ||
| BC008368.1 | - | 4 | 664 | 3355282 | ||

ASUN
Genomic Map