siRNA Details
| Gene Symbol: C14orf105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018168.3 | NP_060638.2 | 3, 4 | 552 | ||
| XM_005267806.1 | XP_005267863.1 | 3, 4 | 545 | |||
| XM_005267811.3 | XP_005267868.1 | 2, 3 | 224 | |||
| XM_005267813.1 | XP_005267870.1 | 3, 4 | 621 | |||
| XM_006720188.2 | XP_006720251.1 | 3, 4 | 545 | |||
| XM_006720189.2 | XP_006720252.1 | 3, 4 | 265 | |||
| GenBank mRNA | AK001512.1 | - | 3, 4 | 541 | ||
| AK223127.1 | - | 3, 4 | 540 | |||

C14orf105
Genomic Map