siRNA Details
| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042367.1 | NP_001035826.1 | 6 | 781 | ||
| XM_017022083.1 | XP_016877572.1 | 5 | 573 | |||
| GenBank mRNA | AK131030.1 | - | 5 | 691 | ||
| BC055412.1 | - | 6 | 762 | |||
| BX113903.1 | - | 4 | 492 | |||

REC114
Genomic Map