siRNA Details
| Gene Symbol: CASC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135281.1 | 3 | 168 | |||
| GenBank mRNA | AK093211.1 | - | 3 | 1764 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASC22| Gene Symbol: CASC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135281.1 | 3 | 168 | |||
| GenBank mRNA | AK093211.1 | - | 3 | 1764 | ||