siRNA Details
| Gene Symbol: LINC00907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046454.1 | 6, 7 | 713 | |||
| GenBank mRNA | BC040725.2 | - | 6, 7 | 692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00907| Gene Symbol: LINC00907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046454.1 | 6, 7 | 713 | |||
| GenBank mRNA | BC040725.2 | - | 6, 7 | 692 | ||