siRNA Details
| Gene Symbol: LINC00907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046454.1 | 5, 6 | 647 | |||
| NR_046455.1 | 4, 5 | 484 | ||||
| GenBank mRNA | AB593073.1 | - | 4, 5 | 484 | ||
| BC040725.2 | - | 5, 6 | 626 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00907| Gene Symbol: LINC00907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046454.1 | 5, 6 | 647 | |||
| NR_046455.1 | 4, 5 | 484 | ||||
| GenBank mRNA | AB593073.1 | - | 4, 5 | 484 | ||
| BC040725.2 | - | 5, 6 | 626 | |||