siRNA Details
| Gene Symbol: HMSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123366.1 | NP_001116838.1 | 2, 3 | 229 | ||
| XM_011525930.2 | XP_011524232.1 | 2, 3 | 324 | |||
| XM_017025710.1 | XP_016881199.1 | 2, 3 | 324 | |||
| GenBank mRNA | BM696260.1 | - | 2, 3 | 229 | ||

HMSD
Genomic Map