siRNA Details
| Gene Symbol: ZNF844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136501.2 | NP_001129973.1 | 3, 4 | 362 | ||
| XM_017026664.1 | XP_016882153.1 | 2, 3 | 208 | |||
| GenBank mRNA | AK098117.1 | - | 3, 4 | 330 | ||
| AK299732.1 | - | 3, 4 | 362 | |||
| AL832297.1 | - | 3, 4 | 348 | |||
| BC125186.1 | - | 3, 4 | 248 | 40122307 | ||

ZNF844
Genomic Map