siRNA Details
| Gene Symbol: ZNF844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136501.2 | NP_001129973.1 | 4 | 1048 | ||
| NR_134326.1 | 3 | 987 | ||||
| XM_017026664.1 | XP_016882153.1 | 3 | 894 | |||
| GenBank mRNA | AK027865.1 | - | 1 | 334 | ||
| AK098117.1 | - | 4 | 1016 | |||
| AK299732.1 | - | 4 | 1048 | |||
| AL832297.1 | - | 4 | 1034 | |||
| BC125185.1 | - | 3 | 876 | |||
| BC125186.1 | - | 4 | 934 | 40122307 | ||

ZNF844
Genomic Map