siRNA Details
| Gene Symbol: SMIM24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136503.1 | NP_001129975.1 | 4 | 399 | ||
| GenBank mRNA | AF161441.1 | - | 3 | 258 | ||
| BC146979.1 | - | 4 | 387 | 9056907 | ||
| BC146984.1 | - | 4 | 387 | 9056912 | ||
| BM769757.1 | - | 4 | 382 | |||

SMIM24
Genomic Map