siRNA Details
| Gene Symbol: C1orf185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136508.1 | NP_001129980.1 | 5 | 559 | ||
| XM_011541282.2 | XP_011539584.1 | 5 | 666 | |||
| GenBank mRNA | AK130995.1 | - | 5 | 565 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf185| Gene Symbol: C1orf185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136508.1 | NP_001129980.1 | 5 | 559 | ||
| XM_011541282.2 | XP_011539584.1 | 5 | 666 | |||
| GenBank mRNA | AK130995.1 | - | 5 | 565 | ||