siRNA Details
| Gene Symbol: LINC01128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047519.1 | 3 | 380 | |||
| GenBank mRNA | AK026873.1 | - | 3 | 272 | ||
| BC102012.1 | - | 3 | 255 | |||
| BC102023.1 | - | 3 | 255 | |||
| BM459718.1 | - | 3 | 253 | |||
| DR005509.1 | - | 3 | 269 | |||
| DR005973.1 | - | 3 | 292 | |||

LINC01128
Genomic Map